AMBH2D6F2B60 Ambeed, Inc.

4-Aminophthalide

  • MDL number MFCD04117950
  • Cas Number : 59434-19-4
  • Molecular Weight : 149.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154-155°C
Purity 98
Storage temp. Keep in dark place,Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 411°C at 760 mmHg
Product Link https://www.ambeed.com/products/59434-19-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/59434-19-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart