AMBH2D6F2F97 Ambeed, Inc.

2-Bromoethylamine hydrobromide

  • MDL number MFCD00012886
  • Cas Number : 2576-47-8
  • Molecular Weight : 204.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/2576-47-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/2576-47-8.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart