AMBH2D6F2FB2 Ambeed, Inc.

O-Benzylhydroxylamine hydrochloride

  • MDL number MFCD00012952
  • Cas Number : 2687-43-6
  • Molecular Weight : 159.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/2687-43-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/2687-43-6.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart