AMBH2D6F2FC5 Ambeed, Inc.

2-Nitro-4-aminodiphenylamine

  • MDL number MFCD00239471
  • Cas Number : 2784-89-6
  • Molecular Weight : 229.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 228-231
bp 417.6�t 760 mmHg
Iupac Name 2-nitro-N~1~-phenyl-1,4-benzenediamine
Inchi Code 1S/C12H11N3O2/c13-9-6-7-11(12(8-9)15(16)17)14-10-4-2-1-3-5-10/h1-8,14H,13H2
Inchi Key WHJNKCNHEVCICH-UHFFFAOYSA-N

Description

Melting Point 228-231
Purity 98%
Flash Point 206
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 417.6�t 760 mmHg
Product Link https://www.ambeed.com/products/2784-89-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/2784-89-6.html
pictograms GHS07
hstatements H315-H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart