AMBH2D6F30CB Ambeed, Inc.

3-((4-Chlorobenzyl)oxy)piperidine hydrochloride

  • MDL number MFCD13561019
  • Cas Number : 1220033-10-2
  • Molecular Weight : 262.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1220033-10-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1220033-10-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart