AMBH2D6F3122 Ambeed, Inc.

2-Bromo-4-hydroxypyridine

  • MDL number MFCD00234040
  • Cas Number : 36953-40-9
  • Molecular Weight : 174

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 425�t 760 mmHg
Product Link https://www.ambeed.com/products/36953-40-9.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/36953-40-9.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart