AMBH2D6F32D6 Ambeed, Inc.

2-((Pyridin-4-ylmethyl)amino)nicotinic acid

  • MDL number MFCD11118784
  • Cas Number : 854382-06-2
  • Molecular Weight : 229.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/854382-06-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/854382-06-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart