AMBH2D6F32F3 Ambeed, Inc.

6-Chloro-N-phenylpyrazin-2-amine

  • MDL number MFCD13562606
  • Cas Number : 642459-03-8
  • Molecular Weight : 205.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/642459-03-8.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/642459-03-8.html
pictograms GHS07
hstatements H302-H319
Availability details for
Quantity
  • Availability
  • Add to Cart