AMBH2D6F34A4 Ambeed, Inc.

3-Bromoisoxazolo[5,4-b]pyridine

  • MDL number MFCD06661866
  • Cas Number : 864872-31-1
  • Molecular Weight : 199.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 95
Flash Point 132°C
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 294.7°C at 760 mmHg
Product Link https://www.ambeed.com/products/864872-31-1.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/864872-31-1.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart