AMBH2D6F34FB Ambeed, Inc.

7-Bromo-2-chlorothieno[3,2-d]pyrimidine

  • MDL number MFCD13193398
  • Cas Number : 1152475-42-7
  • Molecular Weight : 249.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1152475-42-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1152475-42-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart