AMBH2D6F35BE Ambeed, Inc.

4-Methoxycyclohexanone

  • MDL number MFCD01110115
  • Cas Number : 13482-23-0
  • Molecular Weight : 128.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/13482-23-0.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/13482-23-0.html
pictograms GHS07
hstatements H227-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart