AMBH2D6F36AE Ambeed, Inc.

4,4'-Disulfanediylbis(2-aminobutanoic acid)

  • MDL number MFCD00063095
  • Cas Number : 462-10-2
  • Molecular Weight : 268.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/462-10-2.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/462-10-2.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart