AMBH2D6F3746 Ambeed, Inc.

1-N-Boc-2-Methylpropane-1,2-diamine hydrochloride

  • MDL number MFCD11112241
  • Cas Number : 1179361-35-3
  • Molecular Weight : 224.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1179361-35-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1179361-35-3.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart