AMBH2D6F3FEE Ambeed, Inc.

6-Chloro-3-(trifluoromethyl)-1H-pyrazolo[3,4-d]pyrimidine

  • MDL number MFCD18254709
  • Cas Number : 1211590-25-8
  • Molecular Weight : 222.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1211590-25-8.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/1211590-25-8.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart