AMBH2D6F4007 Ambeed, Inc.

2-(Benzo[b]thiophen-2-yl)acetic acid

  • MDL number MFCD09927470
  • Cas Number : 75894-07-4
  • Molecular Weight : 192.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/75894-07-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/75894-07-4.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart