AMBH2D6F40C6 Ambeed, Inc.

3-Ethyl-1-methyl-1H-imidazol-3-ium bromide

  • MDL number MFCD03427610
  • Cas Number : 65039-08-9
  • Molecular Weight : 191.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70-73
Purity 97
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/65039-08-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/65039-08-9.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart