AMBH2D6F40CD Ambeed, Inc.

3-Amino-3-(4-chlorophenyl)-1-propanol

  • MDL number MFCD03412663
  • Cas Number : 68208-26-4
  • Molecular Weight : 185.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/68208-26-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/68208-26-4.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart