AMBH2D6F40E1 Ambeed, Inc.

Fmoc-Gly-Gly-Gly-Gly

  • MDL number MFCD18427731
  • Cas Number : 1001202-16-9
  • Molecular Weight : 468.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1001202-16-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1001202-16-9.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart