AMBH2D6F40EA Ambeed, Inc.

2-Bromo-5-fluoroisonicotinaldehyde

  • MDL number MFCD09800558
  • Cas Number : 1005291-43-9
  • Molecular Weight : 204.00

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1005291-43-9.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1005291-43-9.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart