AMBH2D6F415B Ambeed, Inc.

6-Bromospiro[chroman-2,4'-piperidin]-4-one

  • MDL number MFCD09262414
  • Cas Number : 690632-08-7
  • Molecular Weight : 296.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/690632-08-7.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/690632-08-7.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart