AMBH2D6F4197 Ambeed, Inc.

Cinnoline hydrochloride

  • MDL number MFCD00074974
  • Cas Number : 5949-24-6
  • Molecular Weight : 166.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/5949-24-6.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/5949-24-6.html
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart