AMBH2D6F41A7 Ambeed, Inc.

5,6-Dihydroxy-2,3-dihydro-1H-inden-1-one

  • MDL number MFCD12756505
  • Cas Number : 124702-80-3
  • Molecular Weight : 164.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107-109
Purity 98
Storage temp. Inert atmosphere,Store in freezer, under -20
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/124702-80-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/124702-80-3.html
pictograms GHS07
hstatements H302-H315-H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart