AMBH2D6F4281 Ambeed, Inc.

(2-Aminothiazol-5-yl)methanol

  • MDL number MFCD06203018
  • Cas Number : 131184-73-1
  • Molecular Weight : 130.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102-103°C
Purity 97
Flash Point 159.873°C
Storage temp. Keep in dark place,Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 340.738°C at 760 mmHg
Product Link https://www.ambeed.com/products/131184-73-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/131184-73-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart