AMBH2D6F439C Ambeed, Inc.

2,5-Dibromonicotinonitrile

  • MDL number MFCD13185477
  • Cas Number : 1214340-41-6
  • Molecular Weight : 261.90

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1214340-41-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1214340-41-6.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart