AMBH2D6F43DD Ambeed, Inc.

2-Methoxy-5-(3,4,5-trimethoxyphenethyl)phenol

  • MDL number MFCD06795132
  • Cas Number : 95041-90-0
  • Molecular Weight : 318.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/95041-90-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/95041-90-0.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart