AMBH2D6F4450 Ambeed, Inc.

1-Ethyl-7-nitro-1,2,3,4-tetrahydroquinoline

  • MDL number MFCD07368631
  • Cas Number : 57883-28-0
  • Molecular Weight : 206.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-ethyl-7-nitro-1,2,3,4-tetrahydroquinoline
Inchi Code 1S/C11H14N2O2/c1-2-12-7-3-4-9-5-6-10(13(14)15)8-11(9)12/h5-6,8H,2-4,7H2,1H3
Inchi Key TUCATNRJGJWEKT-UHFFFAOYSA-N

Description

Purity 97%
Flash Point 101.1
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/57883-28-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/57883-28-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart