AMBH2D6F497D Ambeed, Inc.

Methyl 5-amino-1-methyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD10697500
  • Cas Number : 92406-53-6
  • Molecular Weight : 155.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/92406-53-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/92406-53-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart