AMBH2D6F49A7 Ambeed, Inc.

3-Amino-5-fluoro-6-methoxypyridine

  • MDL number MFCD07374960
  • Cas Number : 886372-63-0
  • Molecular Weight : 142.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/886372-63-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/886372-63-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart