AMBH2D6F4B5C Ambeed, Inc.

7-Fluoroisoquinoline

  • MDL number MFCD11226905
  • Cas Number : 1075-12-3
  • Molecular Weight : 147.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Room Temperature
Physical Form Yellow Solid or Liquid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1075-12-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1075-12-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart