AMBH2D6F4B78 Ambeed, Inc.

2-((2-(2-Hydroxyethoxy)-4-nitrophenyl)amino)ethanol

  • MDL number MFCD03265655
  • Cas Number : 59820-43-8
  • Molecular Weight : 242.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/59820-43-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/59820-43-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart