AMBH2D6F4C49 Ambeed, Inc.

Methyl 4-methylbenzo[d]thiazole-6-carboxylate

  • MDL number MFCD12963417
  • Cas Number : 1190320-40-1
  • Molecular Weight : 207.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1190320-40-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1190320-40-1.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart