AMBH2D6F4CBA Ambeed, Inc.

Methyl 2-amino-3-(trifluoromethyl)benzoate

  • MDL number MFCD11226293
  • Cas Number : 64321-95-5
  • Molecular Weight : 219.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/64321-95-5.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/64321-95-5.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart