AMBH2D6F4D53 Ambeed, Inc.

Z-Dab(Boc)-OH

  • MDL number MFCD01632076
  • Cas Number : 49855-91-6
  • Molecular Weight : 352.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (S)-2-(((Benzyloxy)carbonyl)amino)-4-((tert-butoxycarbonyl)amino)butanoic acid
Purity 95
Storage temp. Sealed in dry,2-8
Physical Form Solid or liquid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/49855-91-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/49855-91-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart