AMBH2D6F4D7B Ambeed, Inc.

(1H-Imidazol-4-yl)methanamine hydrochloride

  • MDL number MFCD13176517
  • Cas Number : 66247-84-5
  • Molecular Weight : 133.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (1H-Imidazol-5-yl)methanamine hydrochloride
Purity 97
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/66247-84-5.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/66247-84-5.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart