AMBH2D6F4D8C Ambeed, Inc.

1-Methyl-1,2,3,4-tetrahydroisoquinoline hydrochloride

  • MDL number MFCD17977125
  • Cas Number : 111635-08-6
  • Molecular Weight : 183.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-methyl-1,2,3,4-tetrahydroisoquinoline hydrochloride
Inchi Code 1S/C10H13N.ClH/c1-8-10-5-3-2-4-9(10)6-7-11-8;/h2-5,8,11H,6-7H2,1H3;1H
Inchi Key CFEGVJOLMOPMHU-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/111635-08-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/111635-08-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart