AMBH2D6F4EEB Ambeed, Inc.

Paroxetine

  • MDL number MFCD00869588
  • Cas Number : 61869-08-7
  • Molecular Weight : 329.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114-116°C
Synonym (3S,4R)-3-((Benzo[d][1,3]dioxol-5-yloxy)methyl)-4-(4-fluorophenyl)piperidine
Purity 97
Flash Point 227°C
Storage temp. Keep in dark place,Inert atmosphere,2-8°C
Physical Form liquid
Shipping Temperature Normal
Country of Origin China
Boiling point 451.7°C at 760 mmHg
Product Link https://www.ambeed.com/products/61869-08-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/61869-08-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart