AMBH2D6F4EF3 Ambeed, Inc.

1-Methyl-1H-pyrrolo[2,3-b]pyridine-3-carboxaldehyde

  • MDL number MFCD11054209
  • Cas Number : 171919-36-1
  • Molecular Weight : 160.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/171919-36-1.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/171919-36-1.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart