AMBH2D6F508B Ambeed, Inc.

6-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde

  • MDL number MFCD10697640
  • Cas Number : 383875-59-0
  • Molecular Weight : 180.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/383875-59-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/383875-59-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart