AMBH2D6F50A1 Ambeed, Inc.

(2'-Methyl-[1,1'-biphenyl]-2-yl)diphenylphosphine

  • MDL number MFCD19441232
  • Cas Number : 402822-72-4
  • Molecular Weight : 352.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/402822-72-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/402822-72-4.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart