AMBH2D6F50BD Ambeed, Inc.

2-Amino-2-methylpropanenitrile hydrochloride

  • MDL number MFCD08456816
  • Cas Number : 50846-36-1
  • Molecular Weight : 120.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147-149°C
Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/50846-36-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/50846-36-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart