AMBH2D6F519D Ambeed, Inc.

4-Chloro-5-iodo-2-methyl-7H-pyrrolo[2,3-d]pyrimidine

  • MDL number MFCD09746278
  • Cas Number : 1060815-92-0
  • Molecular Weight : 293.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1060815-92-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1060815-92-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart