AMBH2D6F51ED Ambeed, Inc.

2,5,6-Triaminopyrimidin-4(3H)-one

  • MDL number MFCD00217058
  • Cas Number : 1004-75-7
  • Molecular Weight : 141.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1004-75-7.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1004-75-7.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart