AMBH2D6F51EE Ambeed, Inc.

Lapatinib ditosylate hydrate

  • MDL number MFCD18904381
  • Cas Number : 388082-78-8
  • Molecular Weight : 943.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/388082-78-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/388082-78-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart