AMBH2D6F523B Ambeed, Inc.

6,7-Dihydro-5H-cyclopenta[b]pyridin-7-amine hydrochloride

  • MDL number MFCD12913793
  • Cas Number : 1187930-42-2
  • Molecular Weight : 170.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1187930-42-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1187930-42-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart