AMBH2D6F5285 Ambeed, Inc.

Ethyl 6-chloropicolinate

  • MDL number MFCD09878438
  • Cas Number : 21190-89-6
  • Molecular Weight : 185.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/21190-89-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/21190-89-6.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart