AMBH2D6F56A3 Ambeed, Inc.

1-Phenylpiperidin-4-amine dihydrochloride

  • MDL number MFCD23135281
  • Cas Number : 1082662-38-1
  • Molecular Weight : 249.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 264-265°C
Purity 95
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1082662-38-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1082662-38-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart