AMBH2D6F56E4 Ambeed, Inc.

Ethyl 1-methyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD04967314
  • Cas Number : 88529-79-7
  • Molecular Weight : 154.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Flash Point 98
Storage temp. Sealed in dry,Room Temperature
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 238.5�t 760 mmHg
Product Link https://www.ambeed.com/products/88529-79-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/88529-79-7.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart