AMBH2D6F571D Ambeed, Inc.

3-Hydroxycyclopentanecarboxylic acid

  • MDL number MFCD13659410
  • Cas Number : 101080-22-2
  • Molecular Weight : 130.1418

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Physical Form Colorless to off-white liquid or solid
Shipping Temperature Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/101080-22-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/101080-22-2.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart