AMBH2D6F57D3 Ambeed, Inc.

4-(Difluoromethyl)phenol

  • MDL number MFCD06657967
  • Cas Number : 403648-76-0
  • Molecular Weight : 144.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 5050
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/403648-76-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/403648-76-0.html
pictograms GHS07
hstatements H302-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart