AMBH2D6F581C Ambeed, Inc.

2-Iodo-6-methoxyaniline

  • MDL number MFCD11977372
  • Cas Number : 354574-31-5
  • Molecular Weight : 249.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/354574-31-5.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/354574-31-5.html
pictograms GHS07
hstatements H302-H319
Availability details for
Quantity
  • Availability
  • Add to Cart